Order Entry
ContactUsLinkComponent
Anti-JNK1(Thr183) + JNK2(Thr183) + JNK3(Thr221) Rabbit Polyclonal Antibody
Anti-JNK1(Thr183) + JNK2(Thr183) + JNK3(Thr221) Rabbit Polyclonal Antibody
Catalog #: 77438-274
Supplier:  Bioss
New Product
Anti-JNK1(Thr183) + JNK2(Thr183) + JNK3(Thr221) Rabbit Polyclonal Antibody
Catalog #: 77438-274
Supplier:  Bioss
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    JNK1(Thr183) + JNK2(Thr183) + JNK3(Thr221)
  • Clonality:
    Polyclonal
  • Gene ID:
    5599
  • Host:
    Rabbit
  • Isotype:
    IgG (H, L)
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Conjugation:
    Unconjugated
  • Size:
    100 µl
  • Western blot:
    Yes
  • Storage buffer:
    0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
  • Molecular weight:
    50 kDa
  • Storage temperature:
    Store at –20 °C for 12 months.
  • Concentration:
    1 µg/µl
  • Purification:
    Purified by protein A
  • Packaging:
    Vial

Specifications

About this item

The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is activated by various cell stimuli, and targets specific transcription factors, and thus mediates immediate-early gene expression in response to cell stimuli. The activation of this kinase by tumor-necrosis factor alpha (TNF-alpha) is found to be required for TNF-alpha induced apoptosis. This kinase is also involved in UV radiation induced apoptosis, which is thought to be related to cytochrom c-mediated cell death pathway. Studies of the mouse counterpart of this gene suggested that this kinase play a key role in T cell proliferation, apoptosis and differentiation. Five alternatively spliced transcript variants encoding distinct isoforms have been reported. (provided by RefSeq, Jun 2013).

Type: Primary
Antigen: JNK1(Thr183) + JNK2(Thr183) + JNK3(Thr221)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat

Customers Who Bought This Also Bought