Anti-KCTD9 Rabbit Polyclonal Antibody
77438-098
: Bioss
- Antibody type:Primary
- Antigen name:KCTD9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Gene ID:54793
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:43
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. no.:77438-098
- Packaging:Vial
protein homooligomerization.
Type: Primary
Antigen: KCTD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human