PYREX® Culture Flasks, Long Neck, Corning
Supplier: Corning
Certificates
About this item
The long neck culture flask is basically an Erlenmeyer flask to which a long straight, rimless neck of heavy tubing has been added.
Long neck erlenmeyer flask is ideal for shaker cultures. The longer necks reduce the amount of splashed solution reaching the closure.
Their uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance.
For convenience, these flasks include a marking spot. The approximate neck ext.Ø is 38 mm.
Specifications
- Catalog No:
- 32645-014
- 89090-645
- 89091-452
- 29174-150
- 89090-782
- 29174-152
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey