VWR® Thermotech® Beakers, PTFE
Catalog #: 89026-154
Supplier: VWR International
Log in for availability
About this item
These unique beakers are a combination of a pure PTFE body with a stabilized PTFE carbon base to form a single molding.
- Heatable to 270°C (518°F) without distortion
- Inert
- Autoclavable
Made from prime grade PTFE with FDA and USP Class VI approvals.
Customers Who Bought This Also Bought