VWR® Large-Digit Digital Desk Timer
7×5.1×1.3 cm (2¾×2×½")
Catalog # 82023-844
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
VWR STOPWATCH DIGITAL Catalog #82023-864
Product 82023-864 is similar to 82023-844 but manufacturer and specifications may vary.
Product Details & Documents
Desk timer counts up or down from 99 minutes and 59 seconds.
- Audible buzzer alarm
- Comes with magnetic clip back
Features an audible buzzer alarm, memory for preset value, and a magnetic clip back with a table top stand.
Certifications: CE marked.
Ordering information: Operates on one AAA battery, included.
Specifications
- W×H×D:7×5.1×1.3 cm (2¾×2×½")