Order Entry
ContactUsLinkComponent
Anti-Centaurin beta 2 Rabbit Polyclonal Antibody
Anti-Centaurin beta 2 Rabbit Polyclonal Antibody
Catalog #: 77440-460
Supplier:  Bioss
New Product
Anti-Centaurin beta 2 Rabbit Polyclonal Antibody
Catalog #: 77440-460
Supplier:  Bioss
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Centaurin beta 2
  • Clonality:
    Polyclonal
  • Gene ID:
    23527
  • Host:
    Rabbit
  • Isotype:
    IgG (H, L)
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    100 µl
  • Western blot:
    Yes
  • Storage buffer:
    0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
  • Molecular weight:
    88
  • Storage temperature:
    Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
  • Concentration:
    1 µg/µl
  • Purification:
    Purified by protein A
  • Packaging:
    Vial

Specifications

About this item

The ADP-ribosylation factor (ARF) family of small GTP-binding proteins are involved in vesicular transport regulation and in controlling cytoskeletal organization and cell adhesion. These proteins mainly regulate membrane traffic. ACAP2 is a member of the centaurin GTPase-activating protein (GAP) family, which comprises a subset of ARF regulatory molecules that transduce PI 3-kinase activation into coordinated control of ARF-dependent pathways. ACAP1 and ACAP2 are both widely expressed in peripheral, tubular membranes and usually interact with each other in various tissues. GAP activity of both ACAP1 and ACAP2 is dependent upon phosphatidylinositol 4,5-bisphosphate [PtdIns(4,5)P2]. ACAP2 associates with ARF1 and ARF6. Overexpression of ACAP2 blocks the formation of ARF6-dependent protrusions. K1L is a protein required for growth of the Vaccinia Virus that interacts with the ankyrin repeats of ACAP2.

Type: Primary
Antigen: Centaurin beta 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity:

Customers Who Bought This Also Bought