Order Entry
Canada
Orders LinkContactUsLinkComponent
PYREX® Beakers, Corning

PYREX® Beakers, Corning

3 L (101.4 oz.), Beaker with Handle, 213 mm, 155 mm
Catalog # 89091-352
Supplier:   Corning
Supplier #:  1010
PYREX® Beakers, Corning
Catalog # 89091-352
Supplier:  Corning
PYREX® Beakers, Corning
PYREX® Beakers, Corning

3 L (101.4 oz.), Beaker with Handle, 213 mm, 155 mm

Price
$5,553.00
New Customer Promotional Price
Log in for availability
Case of 8
Product Details & Documents

Made of PYREX® borosilicate glass.

  • Excellent chemical resistance
  • Good chemical resistance

A sturdy, molded glass handle is secured to this 3 L beaker by a stainless steel strap which fits into a recess in the beaker wall. Hot liquids can be handled easily and safely.

Documents

Specifications
  • Ext.Ø:
    155 mm
  • Capacity:
    3 L (101.4 oz.)
  • Description:
    Beaker with Handle
  • Height:
    213 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.