To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:C-RAF/Raf-1
- Clonality:Polyclonal
- Gene ID:5894
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Human,Rat,Mouse
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µl
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:73 kDa
- Storage temperature:Store at −20 °C for 12 months
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Packaging:Vial
Specifications
About this item
cRaf/Raf1 (RAF proto-oncogene serine/threonine-protein kinase) involved in the transduction of mitogenic signals from the cell membrane to the nucleus. Part of the Ras-dependent signaling pathway from receptors to the nucleus. Protects cells from apoptosis mediated by STK3. The Raf family is comprised of three members (aRaf, bRaf, and cRaf) that play a critical role in regulating cell growth and differentiation, and couple growth factor receptor stimulation to nuclear transcription factors via the Ras/mitogen activated protein kinase (MAPK) pathway. cRaf kinase is a small GTPase like kinase of 73 kDa, and is a signal transducer of multiple extracellular stimuli that is regulated by several pathways, and that once activated, phosphorylates MEK which in turn phosphorylates ERK. Raf1 is involved in the transduction of mitogenic signals from the cell membrane to the nucleus. It is part of the Ras dependent signaling pathway from receptors to the nucleus.
Type: Primary
Antigen: C-RAF/Raf-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat