To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:MOXD1
- Clonality:Polyclonal
- Gene ID:26002
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Human,Mouse
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µl
- Western blot:Yes
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:68
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Packaging:Vial
Specifications
About this item
MOXD1 is a 613 amino acid single-pass type I membrane protein of the Endoplasmic reticulum that belongs to the copper type II ascorbate-dependent monooxygenase family. Existing as two alternatively spliced isoforms, MOXD1 is expressed in adult spinal cord, adrenal gland, brain, testis, uterus, lung and kidney, as well as fetal liver and brain. MOXD1 is upregulated during replicative senescence in primary fibroblast and umbilical vein endothelial cell cultures, and uses two copper ions per subunit as a cofactor. MOXD1 contains one DOMON domain, undergoes post-translational N-glycosylation and is encoded by a gene that maps to human chromosome 6. Chromosome 6 contains 170 million base pairs, comprises nearly 6% of the human genome and is associated with early onset intestinal cancer, Porphyria cutanea tarda, Parkinson's disease and Stickler syndrome.
Type: Primary
Antigen: MOXD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse