Anti-COX4/COX IV-1 Rabbit Polyclonal Antibody
Catalog # 77437-892
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:COX4/COX IV-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Size:100 µl
- Gene ID:1327
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Storage temperature:Store at −20 °C for 12 months
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. no.:77437-892
- Packaging:Vial
Specifications
About this item
Cytochrome C Oxidase (COX), which is located in the inner mitochondrial membrane, is the terminal enzyme complex of the mitochondrial electron transport chain. It collects electrons that are transferred from reduced CYTOCHROME C and donates them to molecular oxygen, which is then reduced to water. It is composed of CYTOCHROME A and CYTOCHROME B, two copper atoms, and 13 different protein subunits, three of which are encoded by the mitochondrial DNA and ten others by nuclear DNA.
Type: Primary
Antigen: COX4/COX IV-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat, Bovine