Anti-DNA polymerase epsilon subunit 2 Rabbit Polyclonal Antibody
Catalog # 77440-666
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:DNA polymerase epsilon subunit 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Human,Mouse
- Size:100 µl
- Gene ID:5427
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:59
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. no.:77440-666
- Packaging:Vial
Specifications
About this item
Participates in DNA repair and in chromosomal DNA replication.
Type: Primary
Antigen: DNA polymerase epsilon subunit 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse