Anti-CORO7 Rabbit Polyclonal Antibody
Catalog # 77440-530
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CORO7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Human,Rat,Mouse
- Size:100 µl
- Gene ID:79585
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:101 kDa
- Storage temperature:Store at −20 °C for 12 months
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. no.:77440-530
- Packaging:Vial
Specifications
About this item
Coronins, such as CORO7, constitute an evolutionarily conserved family of WD-repeat actin-binding proteins. CORO7 plays a role in Golgi complex morphology and function (Rybakin et al., 2004, 2006 [PubMed 15327992] [PubMed 16905771]).[supplied by OMIM, Mar 2008].
Type: Primary
Antigen: CORO7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat