Order Entry
ContactUsLinkComponent
Anti-SNAI1+SLUG Rabbit Polyclonal Antibody
Anti-SNAI1+SLUG Rabbit Polyclonal Antibody
Catalog #: 77440-242
Supplier:  Bioss
New Product
Anti-SNAI1+SLUG Rabbit Polyclonal Antibody
Catalog #: 77440-242
Supplier:  Bioss
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SNAI1+SLUG
  • Clonality:
    Polyclonal
  • Gene ID:
    6615
  • Host:
    Rabbit
  • Isotype:
    IgG (H, L)
  • Reactivity:
    Zebrafish,
    Human,
    Rat,
    Mouse
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    100 µl
  • Western blot:
    Yes
  • Storage buffer:
    0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
  • Molecular weight:
    30
  • Storage temperature:
    Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
  • Concentration:
    1 µg/µl
  • Purification:
    Purified by protein A
  • Packaging:
    Vial

Specifications

About this item

SNAI1 is nvolved in induction of the epithelial to mesenchymal transition (EMT), formation and maintenance of embryonic mesoderm, growth arrest, survival and cell migration. Binds to 3 E-boxes of the E-cadherin/CDH1 gene promoter and to the promoters of CLDN7 and KRT8 and, in association with histone demethylase KDM1A which it recruits to the promoters, causes a decrease in dimethylated H3K4 levels and represses transcription. Associates with EGR1 and SP1 to mediate tetradecanoyl phorbol acetate (TPA)-induced up-regulation of CDKN2B, possibly by binding to the CDKN2B promoter region 5'-TCACA-3. In addition, may also activate the CDKN2B promoter by itself. SLUG is transcriptional repressor that modulates both activator-dependent and basal transcription. Involved in the generation and migration of neural crest cells. Plays a role in mediating RAF1-induced transcriptional repression of the TJ protein, occludin (OCLN) and subsequent oncogenic transformation of epithelial cells By similarity. Represses BRCA2 expression by binding to its E2-box-containing silencer and recruiting CTBP1 and HDAC1 in breast cells. In epidermal keratinocytes, binds to the E-box in ITGA3 promoter and represses its transcription. Involved in the regulation of ITGB1 and ITGB4 expression and cell adhesion and proliferation in epidermal keratinocytes. Binds to E-box2 domain of BSG and activates its expression during TGFB1-induced epithelial-mesenchymal transition (EMT) in hepatocytes. Represses E-Cadherin/CDH1 transcription via E-box elements. Involved in osteoblast maturation. Binds to RUNX2 and SOC9 promoters and may act as a positive and negative transcription regulator, respectively, in osteoblasts. Binds to CXCL12 promoter via E-box regions in mesenchymal stem cells and osteoblasts. Plays an essential role in TWIST1-induced EMT and its ability to promote invasion and metastasis.

Type: Primary
Antigen: SNAI1+SLUG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat, Zebrafish

Customers Who Bought This Also Bought