To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:LRP1
- Clonality:Polyclonal
- Gene ID:4035
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Human
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µl
- Western blot:Yes
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:85/515
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Packaging:Vial
Specifications
About this item
Low density lipoprotein receptor-related protein (also known as low density LRP, LRP1, alpha-2-macroglobulin receptor or Apolipoprotein E receptor) is an endocytotic receptor that is involved both in endocytosis and in phagocytosis of apoptotic cells. It is required for early embryonic development, is involved in cellular lipid homeostasis, and may play a role in APP metabolism, kinase-dependent intracellular signalling, neuronal calcium signalling and neurotransmission. Low density LRP also plays a role in the plasma clearance of chylomicron remnants and activated LRPAP1 (alpha-2-macroglobulin), and is involved in the local metabolism of complexes of plasminogen activators and their endogenous ligands. Low density LPR is postulated to be one of the major players in host resistance to HIV. The precursor low density LRP molecule is cleaved post-translationally to form a 85 kDa membrane-spanning subunit (LRP-85) and a 515 kDa large extracellular domain (LRP-515), which remains non-covalently associated with LRP-85. Following cleavage, the intracellular domain (LPRICD) is present in both the cytoplasm and the nucleus.
Type: Primary
Antigen: LRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human