To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CNG3
- Clonality:Polyclonal
- Gene ID:1261
- Host:Rabbit
- Isotype:IgG (H, L)
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µl
- Western blot:Yes
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:79
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Affinity purified by protein A
- Packaging:Vial
Specifications
About this item
Cyclic nucleotide-gated (CNG) cation channels are heteromeric complexes made up of principal alpha and modulatory beta subunits. The alpha subunits consist of CNG1-3 and form functional cation channels by themselves. The beta subunits consist of CNG4-6 and, unlike the alpha subunits, do not form functional channels, but rather modify the properties of channels. CNG channels are essential components of olfactory and visual transduction. In olfactory neurons, CNG2, CNG4.3 and CNG5 form Ca2+ permeable channels, which open and depolarize the cell in response to cAMP. In rod photoreceptors, CNG1 and CNG4.1 combine to form Ca ion permeable channels, which give rise to a current in response to cGMP. CNG3 and CNG6 are expressed in cone receptors and may combine to form a native cGMP-activated channel. CNG channels have been implicated in other areas. CNG1 is also expressed in medium-sized and small-sized arteries, suggesting a role for CNG in the regulation of arterial blood pressure and of blood supply to different regions. CNG1, CNG4.1 and CNG4.2 have been detected in the rat pineal gland. CNG2, CNG4.3 and CNG5 are present in GT1 cell lines and may play a role in the secretion of gonadotropin-releasing hormone.
Type: Primary
Antigen: CNG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: