Trypsinizing Flasks, with Cap, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
About this item
Wheaton-33® low extractable borosilicate glass composition with narrow lip
Deep uniform baffles provide excellent turbulence. Internal dam reduces spill out of untrypsinized particles. Black screw cap with white (styrene-butadiene) rubber liner.
10 Options Available Below
Product Details & Documents
Wheaton-33® low extractable borosilicate glass composition with narrow lip
Deep uniform baffles provide excellent turbulence. Internal dam reduces spill out of untrypsinized particles. Black screw cap with white (styrene-butadiene) rubber liner.
Product Family Options
Product Information
- CapacityCap SizeAvailabilityPrice
- Supplier Number: 355394
- Supplier Number: 355395
- Supplier Number: 355397
- Supplier Number: 355444
- Supplier Number: 355807
- Supplier Number: 355392
- Supplier Number: 355442
- Supplier Number: 355445
- Supplier Number: 355803
- Supplier Number: 355804