Anti-Carbonic anhydrase 3 Rabbit Polyclonal Antibody
77440-200
: Bioss
- Antibody type:Primary
- Antigen name:Carbonic anhydrase 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µl
- Gene ID:761
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:30
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by Protein A
- Cat. no.:77440-200
- Packaging:Vial
Type: Primary
Antigen: Carbonic anhydrase 3
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse, Rat