Order Entry
ContactUsLinkComponent
Anti-TrkA (Tyr490) Rabbit Polyclonal Antibody
Anti-TrkA (Tyr490) Rabbit Polyclonal Antibody
Catalog #: 77438-240
Supplier:  Bioss
New Product
Anti-TrkA (Tyr490) Rabbit Polyclonal Antibody
Catalog #: 77438-240
Supplier:  Bioss
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    TrkA (Tyr490)
  • Clonality:
    Polyclonal
  • Gene ID:
    4914
  • Host:
    Rabbit
  • Isotype:
    IgG (H, L)
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Conjugation:
    Unconjugated
  • Size:
    100 µl
  • Western blot:
    Yes
  • Storage buffer:
    0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
  • Molecular weight:
    90 kDa
  • Storage temperature:
    Store at –20 °C for 12 months.
  • Concentration:
    1 µg/µl
  • Purification:
    Purified by Protein A
  • Packaging:
    Vial

Specifications

About this item

The Trk family of nerve growth factor receptors includes Trk A(also referfed to as Trk A gp140),Trk B and Trk C. The prototype member of this gene family, Trk A, encodes a 140 kDa cell surface receptor , gp140, the expression of which is restricted in vivo to neurons of the sensory spinal and cranial gangliaof neurocrest origin. Nerve growth factor (NGF) stimulates tyrosine phosphorylation of Trk gp 140 in neural cell lines and in embryonic dorsal root ganglia. By comparison, BDNF and to a lesser extent, NT-3, but not NGF, can induce tyrosine phophorylayion of Trk B gp 145. The third member of the Trk receptor family, Trk C incodes a 140 kDa protein, Trk C gp140, that is preferentially expressed in brain tissue and primarily functions as a receptor for NT-3.An additional component of the Trk receptor complex, NGFR p175, binds to neurotrophic factors with low affinity but is required for efficient signaling. NGFR p175 accelerates Trk activation and may recruit downstream dffector molecules to the ligand-bound receptor complex.

Type: Primary
Antigen: TrkA (Tyr490)
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat

Customers Who Bought This Also Bought