Acetone ≥99.9% (dried basis) ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Catalog # CABJAH010-4PC
Supplier: Burdick & Jackson
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle, poly-coated
- Size:4 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 solvent selection guide recommended after discussion
- Cat. no.:CABJAH010-4PC
- Supplier No.:AH010-4PC
Specifications
Specification Test Results
- Aldehyde (as HCHO):0.002 % max.
- Water by Karl Fischer Titration:0.5 % max.
- Color (APHA):10 max.
- GC Analysis (excluding water):99.9 % min.
- Isopropyl Alcohol:0.05 % max.
- Methanol:0.05 % max.
- Residue:3 mg/L max.
- Titrable Acid:0.3 µeq/g max.
- Titrable Base:0.6 µeq/g max.
- UV Cutoff:330 nm max.
Specification Test Results
About this item
:
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56 °C
Melting Pt:–94.8 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:–20 °C
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS07
:GHS02
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765