To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:eIF4A2
- Clonality:Polyclonal
- Gene ID:1974
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Rat
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µl
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:46
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by Protein A
- Packaging:Vial
Specifications
About this item
Translation initiation in eukaryotes necessitates the assembly of an 80S ribosomal complex. Eukaryotic initiation factors (eIFs) are utilized in a sequence of reactions that leads to 80S ribosomal assembly and initiation of translation. Mammalian eukaryotic translation initiation factor 4F (eIF4F) is a protein complex that contains eIF4A, eIF4E and eIF4G, binds mRNA at a 5-cap motif and recruits the 43S ribosomal preinitiation complex to the eligible transcript. Along with eIF4B, the eIF4F complex mediates the unwinding of mRNA secondary structure to facilitate ribosome association. eIF4E specifically interacts with the 5 cap, eIF4A(I,II) are bidirectional RNA helicases, and eIF4G(I,II) are scaffolding proteins which coordinate eIF4E, eIF4A, eIF3 and the 40S ribosome. Human eIF4AI (eIF4A, DDX2A) is a 406 amino acid protein that is 92.7% homologous to mouse eIF4AI. The promoter region of human eIF4A1 contains TATA and CAAT motifs and consensus binding sites to Sp1 and AP2.
Type: Primary
Antigen: eIF4A2
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Rat