Order Entry
ContactUsLinkComponent
Anti-COPG 1/2 Rabbit Polyclonal Antibody
Anti-COPG 1/2 Rabbit Polyclonal Antibody
Catalog #: 77440-526
Supplier:  Bioss
New Product
Anti-COPG 1/2 Rabbit Polyclonal Antibody
Catalog #: 77440-526
Supplier:  Bioss
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    COPG 1/2
  • Clonality:
    Polyclonal
  • Gene ID:
    22820, 26958
  • Host:
    Rabbit
  • Isotype:
    IgG (H, L)
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    100 µl
  • Western blot:
    Yes
  • Storage buffer:
    0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
  • Molecular weight:
    98
  • Storage temperature:
    Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
  • Concentration:
    1 µg/µl
  • Purification:
    Purified by Protein A
  • Packaging:
    Vial

Specifications

About this item

The coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles, which further mediate biosynthetic protein transport from the ER, via the Golgi up to the trans Golgi network. Coatomer complex is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. In mammals, the coatomer can only be recruited by membranes associated to ADP-ribosylation factors (ARFs), which are small GTP-binding proteins; the complex also influences the Golgi structural integrity, as well as the processing, activity, and endocytic recycling of LDL receptors. Required for limiting lipid storage in lipid droplets. Involved in lipid homeostasis by regulating the presence of perilipin family members PLIN2 and PLIN3 at the lipid droplet surface and promoting the association of adipocyte triglyceride lipase (PNPLA2) with the lipid droplet surface to mediate lipolysis (By similarity).

Type: Primary
Antigen: COPG 1/2
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity:

Customers Who Bought This Also Bought