To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:GEP100/IQSEC1
- Clonality:Polyclonal
- Gene ID:9922
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Mouse
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µl
- Western blot:Yes
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:108
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by Protein A
- Packaging:Vial
Specifications
About this item
The ADP-ribosylation factor (ARF) protein family are structurally and functionally conserved members of the Ras superfamily of regulatory GTP-binding proteins. ARFs influence vesicle trafficking and signal transduction in eukaryotic cells. ARF6 plays a role in protein trafficking near the plasma membrane, including receptor recycling, cell adhesion and cell migration. ARF6 colocalizes with the ARF guanine nucleotide-exchange protein (GEP) BRAG2, also designated GEP100. BRAG2 is ubiquitously expressed as two isoforms, BRAG2a and BRAG2b, which can cycle between the cytoplasm and the nucleus. BRAG2, via its interaction with ARF6, is involved in the regulation of cell adhesion by controlling Integrin 1 endocytosis and E-cadherin redistribution. BRAG2 has also been shown to bind directly to Tyr1068/1086-phosphorylated EGFR to activate ARF6, which induces tumor invasion in MCF7 cells. Therefore, BRAG2 may contribute to the metastasis and malignancy of some breast cancer cells.
Type: Primary
Antigen: GEP100/IQSEC1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse