Order Entry
ContactUsLinkComponent
Anti-ANGPTL6 Rabbit Polyclonal Antibody
Anti-ANGPTL6 Rabbit Polyclonal Antibody
Catalog #: 77436-762
Supplier:  Bioss
New Product
Anti-ANGPTL6 Rabbit Polyclonal Antibody
Catalog #: 77436-762
Supplier:  Bioss
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ANGPTL6
  • Clonality:
    Polyclonal
  • Gene ID:
    83854
  • Host:
    Rabbit
  • Isotype:
    IgG (H, L)
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    100 µl
  • Storage buffer:
    0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
  • Molecular weight:
    49
  • Storage temperature:
    Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
  • Concentration:
    1 µg/µl
  • Purification:
    Affinity purified by Protein A
  • Packaging:
    Vial

Specifications

About this item

ngptl6 (angiopoietin-like 6) is a 470 amino acid secreted protein that contains one fibrinogen C-terminal domain and is a member of the angiopoietin-like family. Expressed abundantly in liver and present at lower levels in testis, kidney, heart, brain and lung, Angptl6 plays a role in wound healing and is also thought to promote neovascularization and enhance the chemotactic activity of endothelial cells. Additionally, Angptl6 may be involved in epidermal proliferation, remodeling and regeneration and may be able to counteract expresit increment obesity by . Human Angptl6 shares 74% amino acid identity with its mouse counterpart, suggesting a conserved role between species. The gene encoding Angptl6 maps to human chromosome 19, which is the genetic home for a number of immunoglobulin superfamily members,including the killer cell and leukocyte Ig-like receptors, a number of ICAMs, the CEACAM and PSG family and Fc receptors (FcRs).

Type: Primary
Antigen: ANGPTL6
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity:

Customers Who Bought This Also Bought