Anti-beta 2 Microglobulin Rabbit Polyclonal Antibody
Catalog # 77436-824
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:beta 2 Microglobulin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Gene ID:567
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:13/27
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by Protein A
- Cat. no.:77436-824
- Packaging:Vial
Specifications
About this item
This gene encodes a serum protein found in association with the major histocompatibility complex (MHC) class I heavy chain on the surface of nearly all nucleated cells. The protein has a predominantly beta-pleated sheet structure that can form amyloid fibrils in some pathological conditions. A mutation in this gene has been shown to result in hypercatabolic hypoproteinemia.[provided by RefSeq, Sep 2009].
Type: Primary
Antigen: beta 2 Microglobulin
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human