VWR® QC Timer™ Timer
8.9×5.1×1.3 cm (3½×2×½")
Supplier: VWR International
Jump to:
Product Details & Documents
Ideal quality control countdown and alarm timer combines easy three-button operation with large 1.9 cm (³/₄") digits.
- Traceable®
- Ideal ISO 9000 timer
- Countdown alarm
- Magnet/clip on back
Can be set up to 19 hours, 59 minutes. Alarm sounds for one minute.
: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
: Supplied with three-year AAA alkaline battery.
Specifications
- W×H×D:8.9×5.1×1.3 cm (3½×2×½")