Ammonium chloride 0.100 M (M/10) in aqueous solution analytical reagent
Catalog # RC62716
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly bottle
- Size:16 oz.
- Cat. no.:RC62716
- Supplier No.:R0627000-500A
Specifications
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 999):0.1000 ± 0.0001 N at 20.0 °C
Specification Test Results
About this item
:
:53.49 g/mol
Storage Temperature:Ambient
EINECS:235-186-4
CAS Number:12125-02-9
Formula:NH₄Cl
MDL Number:MFCD00011420