ContactUsLinkComponent
Passive PoE Cable Set for SenseAnywhere AccessPoints: Simplify Installation with Combined Power and Data
Passive PoE Cable Set for SenseAnywhere AccessPoints: Simplify Installation with Combined Power and Data
Catalog # 77441-698
Supplier:  SenseAnywhere
undefined
Passive PoE Cable Set for SenseAnywhere AccessPoints: Simplify Installation with Combined Power and Data
Catalog # 77441-698
Supplier:  SenseAnywhere
Supplier Number:  0630125977095
Specifications
  • Description:
    Passive PoE cable set for SenseAnywhere AccessPoints: Simplify installation with combined power and data
  • Cat. no.:
    77441-698
Specifications
About this item
Optimize the AccessPoint installation with this passive POE splitter. Utilizing a white cable set, it efficiently combines and splits data and power at the switch and AccessPoint, respectively. Designed for setups without POE-enabled switches, it ensures a unified, hassle-free connection with a standard ethernet cable.
  • Passive Power Over Ethernet (POE) cable set: Offers a simple solution to combine power from an adapter and data from a network switch into one ethernet cable
  • Combiner and splitter included: Comes with both a combiner for merging power and data at the switch end and a splitter for separating them at the AccessPoint
  • Ideal for standard network setups: Perfect for setups without a POE-enabled switch, providing flexibility in power and data delivery
  • Enhances installation convenience: Facilitates easy and efficient installation without the need for additional power cables, ensuring a tidy and organized setup
At the location of the switch, use the combiner to combine power from the supplies power adapter and data from the switch. At the AccessPoint, use the splitter to split the data and power before entering the AccessPoint.