Active PoE Splitter, Direct Power and Data Splitting for POE-Enabled SenseAnywhere AccessPoints
Catalog # 77441-696
Supplier: SenseAnywhere
Specifications
- Description:Active PoE splitter: Direct power and data splitting for POE-enabled SenseAnywhere AccessPoints
- Environmentally Preferable:
- Cat. no.:77441-696
Specifications
About this item
Streamline the SenseAnywhere AccessPoint setup with our active POE splitter. Ideal for POE-enabled switch environments, it simplifies connection by splitting both power and data directly, eliminating the need for separate power supplies. A compact, efficient solution for seamless integration.
- Active Power Over Ethernet (POE) splitter: Simplifies the connection process by splitting power and data from a single cable into separate inputs for the AccessPoint
- Designed for POE-Enabled switches: Ideal for environments where the switch already provides both power and data, requiring only splitting at the device end
- Streamlines installation: Eliminates the need for separate power supplies, making installation cleaner and more efficient
- Universal compatibility: Works seamlessly with SenseAnywhere AccessPoints, ensuring reliable and continuous power and data connectivity
When the customer has a POE enabled switch the switch does provide both power and data and users only need to split them using the black active POE splitter before entering the AccessPoint.