Order Entry
Canada
Orders LinkContactUsLinkComponent
Innovating Science Unknown Compositions Samples: Soda Ash

Innovating Science Unknown Compositions Samples: Soda Ash

Unknown set, soda ash, 100 g (sample), 10 samples
Catalog # 470354-896
Supplier:   Ward's Science
Supplier #:  IS5902
Innovating Science Unknown Compositions Samples: Soda Ash
Catalog # 470354-896
Supplier:  Ward's Science
Innovating Science Unknown Compositions Samples: Soda Ash
Innovating Science Unknown Compositions Samples: Soda Ash

Unknown set, soda ash, 100 g (sample), 10 samples

Price
$79.40
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

A series of sodium carbonate (soda ash) samples with a range of known purities.

  • 10×100 g samples with purity concentrations ranging from 20 to 50%.
  • Reference sheet included

Use these in practicing analytical techniques such as gravimetry and acid-base titration.

Specifications
  • Description:
    Unknown set, soda ash
  • Size:
    100 g (sample)
  • No. of uses:
    10 samples

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.