Order Entry
Canada
ContactUsLinkComponent
Innovating Science Unknown Compositions Samples: Potassium Hydrogen Phthalate
Innovating Science Unknown Compositions Samples: Potassium Hydrogen Phthalate
Catalog # 470354-894
Supplier:  Ward's Science
undefined
Innovating Science Unknown Compositions Samples: Potassium Hydrogen Phthalate
Catalog # 470354-894
Supplier:  Ward's Science
Supplier Number:  IS5900

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Unknown set, potassium hydrogen phthalate
  • Size:
    100 g (sample)
  • Cat. no.:
    470354-894
  • No. of Uses:
    10 samples

Specifications

About this item

A series of potassium hydrogen phthalate samples with a range of known purities.

  • 10×100 g samples with purity concentrations ranging from 20 to 65%
  • Reference sheet included

Use these in practicing analytical techniques such as acid-base titration.