Order Entry
Canada
ContactUsLinkComponent
Acetone ≥99.5%, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Flammable
Irritant
Environment
Acetone ≥99.5%, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Catalog # CAJT9009-7
CAS Number:  
MOC:  RegulatedCertificates
undefined
Flammable
Irritant
Environment
Acetone ≥99.5%, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Catalog # CAJT9009-7
Supplier Number:  BAKR9009-07
CAS Number:  
MOC:  RegulatedCertificates

Specifications

  • Pack type:
    Steel pail
  • Size:
    5 gal.
  • Cat. no.:
    CAJT9009-7

Specifications

Specification Test Results

  • Assay ((CH₃)₂CO) (by GC, corrected for water):
    ≥99.5 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤0.001 %
  • Solubility in H₂O:
    Passes Test
  • Titrable Acid (meq/g):
    ≤0.0003
  • Titrable Base (meq/g):
    ≤0.0006
  • Aldehydes (as HCHO):
    ≤0.002 %
  • Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):
    ≤0.05 %
  • Methanol (CH₃OH)(by GC):
    ≤0.05 %
  • Heavy Metals (as Pb):
    ≤0.20 ppm
  • Substances Reducing Permanganate:
    Passes Test
  • Water (H₂O):
    ≤0.5 %
  • Chloride (Cl):
    ≤0.2 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.050 ppm
  • Nickel (Ni):
    ≤0.1 ppm
  • Manganese (Mn):
    ≤0.100 ppm

Specification Test Results

About this item

 :
 :58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
 :Management of Change (MOC) category = R
REACH:01-2119471330-49
 :GHS02
 :GHS07
 :GHS09
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765