

Acetone, low water content ≥99.3%, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
EINECS:200-662-2
UN:1090
ADR:3,II
:Management of Change (MOC) category = NI
Flash Pt:<–20 °C (closed cup)
REACH:01-2119471330-49
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-350 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤330
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Assay ((CH₃)₂CO)(by GC):≥99.3 %
- Residue after Evaporation:≤2.0 ppm
- Titrable Acid (µeq/g):≤0.3
- Titrable Base (µeq/g):≤0.6
- Water (H₂O):≤0.10 %
Specification Test Results
Related Information
Certificates
Search by Lot Number9003 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor High-Purity Solvents and Reagents Avantor Portfolio of Chemicals
Product Literature
Frequently Bought Together