ContactUsLinkComponent
Nalgene® Drop-Dispenser Bottle, Teflon® FEP, Thermo Scientific
Nalgene® Drop-Dispenser Bottle, Teflon® FEP, Thermo Scientific
Catalog # 16354-320
undefined
Nalgene® Drop-Dispenser Bottle, Teflon® FEP, Thermo Scientific
Catalog # 16354-320
Supplier Number:  2414-0030
Specifications
  • Capacity:
    30 ml (1 oz.)
  • Cat. no.:
    16354-320
  • Supplier No.:
    2414-0030
Specifications
About this item
Made of Teflon® FEP and ETFE dropping closure and cap. Transparent bottle can be used with virtually any chemical. Contamination-free dispensing. Repeatedly autoclavable with tip uncapped.
Convenient one-hand operationjust squeeze bottle to dispense. Dispenses one drop at a time.