KIMAX® Baffled Culture Flasks, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
Designed for use with rotary and reciprocating shakers where baffle indents provide greater turbulence to improve oxygen transfer rates or aeration
Flask has a neck with a plain fire-polished finish suitable for a metal cap, plastic cap, foam plug or cotton plug.
Ordering information: Closures are available separately.