ContactUsLinkComponent
KIMAX® Heavy-Duty Beakers, Low Form, Double Scale, Borosilicate Glass
KIMAX® Heavy-Duty Beakers, Low Form, Double Scale, Borosilicate Glass
Catalog # 89001-082
Supplier #: 14005-2000
undefined
KIMAX® Heavy-Duty Beakers, Low Form, Double Scale, Borosilicate Glass
Catalog # 89001-082
Supplier Number:  14005-250
Supplier #: 14005-2000
Log in for availability
Select an Option
200 ml
Select an Option
200 - 1800 ml
Select an Option
2 L (68 oz.)
Select an Option
89001-082
Select an Option
14005-2000
About this item
Beakers feature thick, uniform walls throughout and an extra-thick, evenly tooled top rim
Spout is designed for optimum performance. Heavy construction is intended to prolong life expectancy with increased and more rigorous usage. All sizes have a durable matte finish marking area and a white graduated scale.
Certifications: Designed from ASTM Specification E960, Type II requirements.
Frequently Bought Together