VWR®, Griffin Low Form Beakers with Double-Capacity Scale, Borosilicate Glass
Catalog #: 89000-204
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: VW14000-250
Log in for availability
About this item
Beakers feature a thick, slightly flared, beaded top with a spout designed to have excellent pouring characteristics.
- Thick, slightly flared, beaded top with a spout designed to have excellent pouring characteristics
- Uniform walls and are made of Type I, Class A borosilicate glass
- Feature a durable matte finish marking area suitable for use with an ordinary pencil
They have uniform walls and are made of Type I, Class A borosilicate glass. Beakers 20 mL and larger are printed with an easy-to-read white graduated scale for approximate measurement. All beakers feature a durable matte finish marking area suitable for use with an ordinary pencil.
Customers Who Bought This Also Bought