Nalgene® Graduated Beakers with Handle, High-Density Polyethylene, Thermo Scientific
Catalog #: 13916-955
Supplier: Thermo Fisher Scientific
Supplier #: 1220-1250
Log in for availability
About this item
Made from high-density polyethylene with extra heavy wall
- Graduated in both ounces and milliliters
- Large rectangular handle
Especially useful in handling acids, alkalies, and photographic chemicals.
Customers Who Bought This Also Bought