Tube Rack, Coated Wire, 16 to 18 mm, 15-Place
Catalog # 77395-376
Supplier: Heathrow Scientific
New Product
Specifications
- Color:Blue
- Material:HDPE/carbon steel
- Cat. no.:77395-376
- For Tubes:Tube array: 3×5; wells: 15; Well Ø: 16 - 18 mm
Specifications
About this item
Chemically resistant HDPE coated wire racks.
- Holds all brands and types of conical, round or flat bottom, glass or plastic, tubes and/or bottles
- Square wells and three deck levels help keep tubes secure
The HDPE coated wire rack is durable for transport or sample handling. It is also chemically resistant and suitable for conical, round or flat-bottom, or plastic tubes and bottles.
Frequently Bought Together