Order Entry
Canada
Orders LinkContactUsLinkComponent
Low Temp Microscope Slide Box, 25-Place

Low Temp Microscope Slide Box, 25-Place

PC, 25-place (26×76 mm and 1×3 in slide size), Blue
Catalog # 77395-314
Supplier:   VWR International
Low Temp Microscope Slide Box, 25-Place
Catalog # 77395-314
Supplier:  VWR International
Low Temp Microscope Slide Box, 25-Place
Low Temp Microscope Slide Box, 25-Place

PC, 25-place (26×76 mm and 1×3 in slide size), Blue

Price
$19.26
New Customer Promotional Price
Log in for availability
Each Case of 72
Product Details & Documents

Store in ultra-low temperatures with confidence.

  • Holds standard slides (76×26 mm and 3×1")
  • Durable Construction
  • Made of polycarbonate to store in ultra-low temperatures
  • Uniform grooved slide slots separate slides preventing slide contact
  • Molded feet enable secure stacking for easy transport and storage
  • Durable Construction
  • Low temperature resistant numbered foam lining corresponds with identification sheet
  • Organize with 'Project' and 'Date' pre-molded labeling area

The low temperature microscope slide box protects your slides in ultra-low temperature ranges down to −80 °C.

Specifications
  • Color:
    Blue
  • Material:
    PC
  • For use with:
    25-place (26×76 mm and 1×3 in slide size)