VITLAB® Graduated Pitchers, SAN, BrandTech
Catalog #: 83008-744
Supplier: Brandtech
Supplier #: 44291
Log in for availability
About this item
Clear SAN (styrene acrylonitrile copolymer) pitchers with molded graduations are a lightweight, safer alternative to glass in many applications.
- Glass-clear pitchers have contoured handles for easy lifting
- Durable molded graduations for long measuring life
- Pouring lip reduces drips
- SAN has a maximum temperature for use of 70 ˚C
Enhanced graduations are provided at major intervals for easy volume estimations.
Customers Who Bought This Also Bought