ContactUsLinkComponent
Bottle Rack, Coated Wire, 55 to 60 mm, 6-Place
Bottle Rack, Coated Wire, 55 to 60 mm, 6-Place
Catalog # 77395-378
undefined
Bottle Rack, Coated Wire, 55 to 60 mm, 6-Place
Catalog # 77395-378
Supplier Number:  HS23224
Specifications
  • Color:
    Blue
  • Material:
    HDPE/carbon steel
  • Environmentally Preferable:
  • Cat. no.:
    77395-378
  • For Vials:
    Tube array: 2×3; wells: 6; well Ø: 55 - 60 mm
Specifications
About this item
Chemically resistant HDPE coated wire racks.
  • Holds all brands and types of conical, round or flat bottom, glass or plastic, tubes and/or bottles
  • Square wells and three deck levels help keep tubes secure
The HDPE coated wire rack is durable for transport or sample handling. It is also chemically resistant and suitable for conical, round or flat-bottom, or plastic tubes and bottles.