Order Entry
Canada
ContactUsLinkComponent
5-(Ethylthio)-1H-tetrazole (ETT) 0.45 M in acetonitrile, Novabiochem® activator reagent, Sigma-Aldrich®
Flammable
Corrosive
Irritant
5-(Ethylthio)-1H-tetrazole (ETT) 0.45 M in acetonitrile, Novabiochem® activator reagent, Sigma-Aldrich®
Catalog # CABI0158-NP20
Supplier:  MilliporeSigma
CAS Number:  
Flammable
Corrosive
Irritant
5-(Ethylthio)-1H-tetrazole (ETT) 0.45 M in acetonitrile, Novabiochem® activator reagent, Sigma-Aldrich®
Catalog # CABI0158-NP20
Supplier:  MilliporeSigma
Supplier Number:  BI0158-NP20
CAS Number:  

Specifications

  • Pack type:
    Plastic barrel
  • Size:
    20 L
  • Cat. no.:
    CABI0158-NP20
  • Supplier No.:
    BI0158-NP20

Specifications

About this item

 :
ADR:3.8,II
 :130.17 g/mol
UN:2924
 :GHS02
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:89797-68-2
Formula:C₃H₆N₄S
MDL Number:MFCD00068733