Orders LinkContactUsLinkComponent
Xcelite® Screwdriver Sets, Apex
Xcelite® Screwdriver Sets, Apex
Round Blade Screwdriver Set
Catalog # 37699-716
Supplier:   Cooper Tools
Supplier #:  SDR11N
Xcelite® Screwdriver Sets, Apex
Catalog # 37699-716
Supplier:  Cooper Tools
Xcelite® Screwdriver Sets, Apex
Xcelite® Screwdriver Sets, Apex
Round Blade Screwdriver Set
Price
$363.78
Log in for availability
Each
Product Details & Documents
Round blade screwdriver set 37699-716 consists of five round blade screwdrivers for slotted screws of the following blade diameters: 1/8", 5/32", 3/16", 1/4", and 5/16".
Specifications
  • Description:
    Round Blade Screwdriver Set