Eisco Micropipette Tips, 10000 µl
10000 µl, -, 15 cm, Eisco VVM12, VVM13, 100/Pack
Catalog # 470355-094
Supplier: Wards
Supplier #: CH5057
Jump to:
Product Details & Documents
10000 µl capacity non-sterile micropipette tips. Free from RNase, DNase, and endotoxins.
- 100 pack of micropipette tips in sealed bag
Fits Eisco variable volume micropipettes with a 10000 µl top range (models VVM112 and VVM13). Each micropipette tip measures 15 cm long and is calibrated for delivery.
Specifications
- Volume:10000 µl
- Length:15 cm
- Packed:100/Pack
- Sterile:-
- For use with:Eisco VVM12, VVM13