About this item
Enhanced GFP (eGFP) is a modified version of GFP that exhibits even stronger fluorescence for easier detection.
- Protein class: Beta-barrel; Autofluorescent protein
- Sequence: Sections named from N-Ter to C-Tery: GST-tag, Thrombin cleavage site, HRV3C (PreScission) cleavage site, eGFP, Spacer, Rho1D4-tag
- Affinity tags: GST-tag (N-terminus), Rho1D4-tag (C-terminus)
- Excitation maximum: 488 nm
- Fluorescence maximum: 509 nm
Purified eGFP can find multiple uses in scientific research. Often as positive controls for protein purifications or western blots. Therefore, we offer eGFP tagged with the most commonly used affinity tags.
We offer the following affinity-tag combinations:
N-Terminal His-tag
N-Terminal GST-tag and C-Terminal Rho1D4-tag
N-Terminal GST-tag and C-Terminal DYKDDDDK-tag
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.