Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Recombinant MSP2N2 (from <i>E. coli</i>)

Human Recombinant MSP2N2 (from E. coli)

Supplier:   Cube Biotech
Human Recombinant MSP2N2 (from <i>E. coli</i>)
Supplier:  Cube Biotech
Certificates
Human Recombinant MSP2N2 (from <i>E. coli</i>)

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Membrane scaffold protein, human origin.

  • Extinction coefficient (in water) ε280: 36900 M⁻¹cm⁻¹ (Human origin), request datasheet (Mouse origin)
  • Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
  • Affinity tag: none

Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP2N2 creates nanodiscs of ~17 nm in diameter.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 77392-682
  • 77392-684
  • Protein/peptide name:
  • MSP2N2
  • MSP2N2
  • Protein synonyms:
  • Membrane Scaffold Protein 2N2
  • Membrane Scaffold Protein 2N2
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • Sequence:
  • G STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ GT PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ
  • G STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ GT PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ
  • Amino acid number:
  • 371 (Human origin)
  • 371 (Human origin)
  • Purity:
  • >90% (Determined by SDS-PAGE)
  • >90% (Determined by SDS-PAGE)
  • Molecular weight:
  • 4305 kDa (Human origin)
  • 4305 kDa (Human origin)
  • Storage conditions:
  • Storage of lyophilized copolymer: −20 °C for several years; Storage of reconstituted protein: 2...8 °C for several day
  • Storage of lyophilized copolymer: −20 °C for several years; Storage of reconstituted protein: 2...8 °C for several day
  • Shipping temperature:
  • RT
  • RT

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 2 mg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $893.10
    $893.10
    Each(1/Itm/Bx)
       $893.10
    Minimum order quantity is 1

    Supplier Number: 26182
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77392-682
    Protein/peptide name
    MSP2N2
    Protein synonyms
    Membrane Scaffold Protein 2N2
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Sequence
    G STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ GT PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ
    Amino acid number
    371 (Human origin)
    Purity
    >90% (Determined by SDS-PAGE)
    Molecular weight
    4305 kDa (Human origin)
    Storage conditions
    Storage of lyophilized copolymer: −20 °C for several years; Storage of reconstituted protein: 2...8 °C for several day
    Shipping temperature
    RT
  • 10 mg
    Log in for availability
    Log in for availability
    Pack of 1 (1/Itm/Bx)
    Pack of 1 (1/Itm/Bx) - $3,970.08
    $3,970.08
    Pack of 1 (1/Itm/Bx)
       $3,970.08
    Minimum order quantity is 1

    Supplier Number: 26186
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77392-684
    Protein/peptide name
    MSP2N2
    Protein synonyms
    Membrane Scaffold Protein 2N2
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Sequence
    G STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ GT PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ
    Amino acid number
    371 (Human origin)
    Purity
    >90% (Determined by SDS-PAGE)
    Molecular weight
    4305 kDa (Human origin)
    Storage conditions
    Storage of lyophilized copolymer: −20 °C for several years; Storage of reconstituted protein: 2...8 °C for several day
    Shipping temperature
    RT
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.