Human Recombinant MSP1D1 (from E. coli)
Supplier: CUBE BIOTECH
Certificates
About this item
Membrane scaffold protein, human origin.
- Extinction coefficient (in water) ε280: 25,440 M⁻¹cm⁻¹ (Human origin), 20,970 M⁻¹cm-⁻¹ (Mouse origin)
- Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
- Affinity tag: none
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences to stabilize membrane proteins in a lipid environment after removing the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 (His-tagged) creates nanodiscs of 9 to 10 nm in diameter.
This product is available with different species as its background. This is important for e.g. for secondary antibodies of a western blot.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together