Human Recombinant MSP1D1 dH5-His (from E. coli)
Membrane scaffold protein, human origin.
- Extinction coefficient (in water) ε280: 19,940 M⁻¹cm⁻¹
- Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 dh5 (his-tagged) creates nanodiscs of 7 to 8 nm in diameter. This product contains a his-tag, keep that in mind when choosing the affinity tag for your membrane protein.
This product is available with different species as its background. This is important for e.g. for secondary antibodies of a western blot.
- Source:
- Species:
- Environmentally Preferable:
Frequently Bought Together