Order Entry
Canada
Orders LinkContactUsLinkComponent
 

PureCube Carboxy MagBeads

Supplier:   Cube Biotech
PureCube Carboxy MagBeads
Supplier:  Cube Biotech
Certificates
PureCube Carboxy MagBeads
PureCube Carboxy MagBeads

1 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

PureCube amine carboxy agarose MagBeads are your best option to couple biomolecules that carry free-standing Amine (NH₂) groups.

  • Usage: Coupling biomolecules to active Carboxy Groups
  • Specificity: Exposed Amine (NH₂) groups on biomolecule
  • Amine group density: Higher than 15 µmol/ml
  • Bead: Ligand chemically active Carboxy groups
  • Bead size: 30 μm
  • Filling quantity: Delivered as a 25% suspension
  • Matrix: 6% cross-linked agarose
  • Short term storage: In equilibration buffer (see protocol)
  • Long term storage: 100 mM sodium hydrogen carbonate, 0.02% sodium azide, pH 7.5 at 4 °C
  • or 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
  • Shelf life: If properly stored - three years upon delivery, unopened
  • Chemical structure: Carboxy structure

The resulting peptide bond between the Carboxy MagBeads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube Carboxy MagBeads are also available as agarose resin if you prefer this purification method or as XL MagBeads if your downstream application involves a very viscous cell media.

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 ml
    Discontinued
    Supplier Number: 50201
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 5 ml
    Discontinued
    Supplier Number: 50205
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 25 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $4,142.61
    $4,142.61
    Each(1/Itm/Bx)
       $4,142.61
     

    Supplier Number: 50225
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 4×25 ml
    Discontinued
    Supplier Number: 50290
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.