Order Entry
Canada
ContactUsLinkComponent
Synthetic Nanodisc Screening Kit MAXI
Synthetic Nanodisc Screening Kit MAXI
Catalog # 77392-568
Supplier:  CUBE BIOTECH
CAS Number:  
undefined
Synthetic Nanodisc Screening Kit MAXI
Catalog # 77392-568
Supplier:  CUBE BIOTECH
Supplier Number:  18295
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    23×2×50 mg
  • Environmentally Preferable:
  • Cat. no.:
    77392-568

Specifications

About this item

Synthetic copolymers are capable of solubilizing and stabilizing membrane proteins without the need for detergents. They have the unique capacity to extract the membrane protein of interest while maintaining a near-native environment.

The versatility of membrane proteins and their composition mirrors the intricate nature of the cell membrane. As a result, it can be a challenge to predict with absolute certainty the optimal synthetic polymer for the solubilization and subsequent stabilization of your membrane protein of interest. Therefore, we have combined our most prominent SMALP, DIBMA, AASTY, and Amphipol products into this practical Synthetic Nanodisc Screening Set MAXI to help you find the correct product.

All 23 products are buffered in HEPES to provide maximal comparability between the individual screening assays.

Delivery: Shipping temperature: Ambient temperature; Storage of lyophilized copolymer: –20 °C for several years; Storage of dissolved copolymer: 2 to 8 °C for several day.